Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015767.3 | NP_056582.1 | 1, 2 | 210 | ||
GenBank mRNA | AF218416.1 | - | 1, 2 | 198 | ||
AK004882.1 | - | 1, 2 | 191 | |||
AK050374.1 | - | 1, 2 | 211 | |||
BC099868.1 | - | 1, 2 | 209 | 40040166 | ||
BC099869.1 | - | 1, 2 | 211 | 40040167 | ||
BC099870.1 | - | 1, 2 | 211 | 40040169 | ||
BC099871.1 | - | 1, 2 | 210 |