Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015773.2 | NP_056588.1 | 6 | 843 | ||
GenBank mRNA | AF173866.1 | - | 6 | 780 | ||
AK005732.1 | - | 6 | 791 | |||
AK054199.1 | - | 5 | 879 | |||
BC090660.1 | - | 6 | 786 | 30673350 | ||
BC100455.1 | - | 6 | 793 | 30919921 |