Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015791.3 | NP_056606.2 | 4, 5 | 816 | ||
GenBank mRNA | AF176527.1 | - | 4, 5 | 823 | ||
AK012401.1 | - | 4, 5 | 836 | |||
AK029989.1 | - | 4, 5 | 817 | |||
AK077595.1 | - | 4, 5 | 838 | |||
AK078111.1 | - | 4, 5 | 836 | |||
AK147748.1 | - | 4, 5 | 833 | |||
AK160145.1 | - | 4, 5 | 799 | |||
BC034854.1 | - | 4, 5 | 748 | 3468704 |