Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163704.1 | NP_001157176.1 | 5 | 892 | ||
NM_001163705.1 | NP_001157177.1 | 5 | 676 | |||
NM_001163706.1 | NP_001157178.1 | 5 | 668 | |||
NM_001163707.1 | NP_001157179.1 | 5 | 703 | |||
NM_015797.4 | NP_056612.1 | 5 | 1418 | |||
GenBank mRNA | AF176526.1 | - | 5 | 791 | ||
AK002840.1 | - | 5 | 630 | |||
AK144715.1 | - | 5 | 829 | |||
AK150612.1 | - | 5 | 860 | |||
AK152888.1 | - | 5 | 742 | |||
AK165867.1 | - | 5 | 1403 | |||
AK170743.1 | - | 5 | 669 | |||
AK171483.1 | - | 5 | 669 | |||
AK178080.1 | - | 11 | ||||
BC017512.1 | - | 5 | 688 | 4485218 |