Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015830.1 | NP_056645.1 | 4 | 1970 | ||
GenBank mRNA | AK079422.1 | - | 3 | 2062 | ||
AK143711.1 | - | 6 | 2609 | |||
AK148209.1 | - | 2 | 229 | |||
AK170543.1 | - | 5 | 2204 | |||
BC058094.1 | - | 5 | 2228 | 6824892 |