Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015830.1 | NP_056645.1 | 6 | 2358 | ||
GenBank mRNA | AK079422.1 | - | 5 | 2450 | ||
AK143711.1 | - | 8 | 2997 | |||
AK148209.1 | - | 4 | 617 | |||
AK170543.1 | - | 7 | 2592 | |||
BC058094.1 | - | 7 | 2616 | 6824892 |