Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016698.2 | NP_057907.2 | 10, 11 | 2136 | ||
GenBank mRNA | AB026621.1 | - | 10, 11 | 2135 | ||
AK004579.1 | - | 10, 11 | 2134 | |||
AK034953.1 | - | 10, 11 | 2135 | |||
AK129106.1 | - | 10, 11 | 2124 | |||
AK146728.1 | - | 10, 11 | 2136 | |||
AK149972.1 | - | 10, 11 | 2137 | |||
AK156502.1 | - | 10, 11 | 2136 | |||
AK165617.1 | - | 10, 11 | 2136 | |||
AK168523.1 | - | 10, 11 | 2131 | |||
AK172065.1 | - | 10, 11 | 2134 | |||
AK194865.1 | - | 1, 2 | 107 | |||
BC010342.1 | - | 10, 11 | 1845 | 3602969 |