Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162870.1 | NP_001156342.1 | 2 | 492 | ||
NM_016962.2 | NP_058658.2 | 2 | 489 | |||
GenBank mRNA | AJ246002.1 | - | 2 | 114 | ||
AK007793.1 | - | 2 | 269 | |||
AK044900.1 | - | 2 | 344 | |||
AK129282.1 | - | 2 | 466 | |||
BC046286.1 | - | 2 | 489 | 6441742 |