Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Keap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110305.1 | NP_001103775.1 | 3 | 1337 | ||
NM_001110306.1 | NP_001103776.1 | 3 | 1222 | |||
NM_001110307.1 | NP_001103777.1 | 3 | 1054 | |||
NM_016679.4 | NP_057888.1 | 2 | 2373 | |||
GenBank mRNA | AB020063.1 | - | 3 | 826 | ||
AK004738.1 | - | 3 | 1054 | |||
AK039549.1 | - | 3 | 993 | |||
AK046178.1 | - | 3 | 938 | |||
AK076234.1 | - | 2 | 1859 | |||
AK129061.1 | - | 2 | 808 | |||
AK136319.1 | - | 1 | 58 | |||
AK150437.1 | - | 3 | 958 | |||
AK150485.1 | - | 3 | 825 | |||
AK152142.1 | - | 3 | 958 | |||
AK154430.1 | - | 3 | 869 | |||
AK155507.1 | - | 3 | 869 | |||
AK159858.1 | - | 3 | 823 | |||
BC006729.1 | - | 3 | 835 | |||
BC023945.1 | - | 3 | 1087 | |||
BC055732.1 | - | 2 | 811 | 6404252 |