Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neu3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016720.2 | NP_057929.1 | 3 | 647 | ||
GenBank mRNA | AB026842.1 | - | 3 | 646 | ||
AK050428.1 | - | 3 | 525 | |||
AK134405.1 | - | 3 | 604 | |||
BC116892.1 | - | 3 | 436 | 40126211 | ||
BC116924.1 | - | 3 | 436 | 40126243 |