Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016758.3 | NP_058038.2 | 1, 3 | 215 | ||
GenBank mRNA | AK154944.1 | - | 1, 3 | 140 | ||
BC030321.1 | - | 1, 3 | 187 | 5374531 | ||
U85055.1 | - | 1, 3 | 55 |