Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016891.3 | NP_058587.1 | 1 | 89 | ||
GenBank mRNA | AB021743.1 | - | 1 | 37 | ||
AK054239.1 | - | 1 | 49 | |||
AK078135.1 | - | 1 | 68 | |||
AK088776.1 | - | 1 | 61 | |||
BC006606.1 | - | 1 | 89 | 3157293 | ||
BC052678.1 | - | 1 | 85 | 30062580 |