Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290824.1 | NP_001277753.1 | 9 | 1212 | ||
NM_153387.3 | NP_700436.1 | 9 | 1212 | |||
GenBank mRNA | AK016563.1 | - | 9 | 1255 | ||
AK038921.1 | - | 9 | 1171 | |||
AK045305.1 | - | 9 | 1105 | |||
AK046132.1 | - | 9 | 1142 | |||
AK050250.1 | - | 9 | 1213 | |||
AK051517.1 | - | 9 | 1113 | |||
AK152984.1 | - | 9 | 1171 | |||
AK153171.1 | - | 9 | 1171 | |||
AK181112.1 | - | 1 | 50 | |||
BC029106.1 | - | 9 | 1176 | 3711922 |