Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057172.3 | NP_476513.2 | 17 | 1740 | ||
GenBank mRNA | AK028418.1 | - | 17 | 1742 | ||
AK138011.1 | - | 17 | 1685 | |||
AK143419.1 | - | 1 | 1396 | |||
AK160815.1 | - | 17 | 1678 | |||
AK166066.1 | - | 17 | 1685 | |||
AK167730.1 | - | 17 | 1720 | |||
BC014763.1 | - | 17 | 1703 | 4511225 | ||
BC023780.1 | - | 17 | 1721 | 5347067 |