Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057172.3 | NP_476513.2 | 12, 13 | 1159 | ||
GenBank mRNA | AK028418.1 | - | 12, 13 | 1160 | ||
AK032517.1 | - | 12, 13 | 1126 | |||
AK138011.1 | - | 12, 13 | 1104 | |||
AK160815.1 | - | 12, 13 | 1097 | |||
AK166066.1 | - | 12, 13 | 1104 | |||
AK167730.1 | - | 12, 13 | 1139 | |||
BC014763.1 | - | 12, 13 | 1122 | 4511225 | ||
BC023780.1 | - | 12, 13 | 1140 | 5347067 |