Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178607.4 | NP_848722.1 | 5 | 439 | ||
GenBank mRNA | AK015699.1 | - | 6 | 499 | ||
AK048813.1 | - | 5 | 397 | |||
AK083665.1 | - | 5 | 429 | |||
AK089207.1 | - | 5 | 426 | |||
AK154093.1 | - | 5 | 376 | |||
AK165427.1 | - | 5 | 397 | |||
BC096529.1 | - | 5 | 372 | 6406985 |