Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033128.3 | NP_001028300.1 | 15 | 1528 | ||
GenBank mRNA | AK037753.1 | - | 15 | 1514 | ||
AK045850.1 | - | 15 | 1526 | |||
AK163815.1 | - | 6 | 696 | |||
BC076577.1 | - | 15 | 1517 |