Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026504.2 | NP_080780.1 | 2, 3 | 373 | ||
GenBank mRNA | AK007496.1 | - | 2, 3 | 374 | ||
AK009825.1 | - | 2, 3 | 369 | |||
AK014348.1 | - | 2, 3 | 385 | |||
AK045993.1 | - | 2, 3 | 380 | |||
AK050334.1 | - | 2, 3 | 380 | |||
AK217127.1 | - | 1, 2 | 142 | |||
BC052345.1 | - | 2, 3 | 380 | 6585618 |