Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026792.3 | NP_081068.1 | 8 | 1356 | ||
GenBank mRNA | AK003649.1 | - | 8 | 1100 | ||
AK082137.1 | - | 8 | 1131 | |||
AK089885.1 | - | 8 | 1159 | |||
AK152709.1 | - | 8 | 1099 | |||
AK152889.1 | - | 8 | 1103 | |||
AK155378.1 | - | 9 | 1305 | |||
AK170093.1 | - | 8 | 1100 | |||
AK213021.1 | - | 2 | 291 | |||
AY161042.1 | - | 8 | 1333 | |||
BC031987.1 | - | 8 | 1081 | 3982362 | ||
BC062903.1 | - | 8 | 1360 |