Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172762.2 | NP_766350.2 | 8, 9 | 907 | ||
GenBank mRNA | AK016057.1 | - | 7, 8 | 839 | ||
AK078099.1 | - | 8, 9 | 900 | |||
BC064035.1 | - | 8, 9 | 886 | |||
BC141278.1 | - | 8, 9 | 896 | 9055890 |