Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027897.4 | NP_082173.3 | 9 | 1110 | ||
GenBank mRNA | AK004849.1 | - | 9 | 1033 | ||
AK050214.1 | - | 9 | 1024 | |||
BC100314.1 | - | 9 | 1017 | 30848276 |