Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026538.3 | NP_080814.1 | 2 | 185 | ||
GenBank mRNA | AK011136.1 | - | 2 | 195 | ||
AK076047.1 | - | 2 | 203 | |||
AK076975.1 | - | 2 | 185 | |||
AK077514.1 | - | 2 | 198 | |||
AK145628.1 | - | 2 | 226 | |||
AK159793.1 | - | 2 | 189 | |||
AK169654.1 | - | 2 | 200 | |||
AK191115.1 | - | 1 | 84 | |||
BC018291.1 | - | 2 | 179 | 4190167 |