Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026538.3 | NP_080814.1 | 2, 3 | 232 | ||
GenBank mRNA | AK011136.1 | - | 2, 3 | 242 | ||
AK076047.1 | - | 2, 3 | 250 | |||
AK076975.1 | - | 2, 3 | 232 | |||
AK077514.1 | - | 2, 3 | 245 | |||
AK145628.1 | - | 2, 3 | 273 | |||
AK159793.1 | - | 2, 3 | 236 | |||
AK169654.1 | - | 2, 3 | 247 | |||
AK191115.1 | - | 1, 2 | 131 | |||
BC018291.1 | - | 2, 3 | 226 | 4190167 |