Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144523.2 | NP_653106.1 | 2, 3 | 1077 | ||
GenBank mRNA | AK031232.1 | - | 2, 3 | 1077 | ||
AK149106.1 | - | 2, 3 | 1008 | |||
AK154951.1 | - | 2, 3 | 1003 | |||
BC006964.1 | - | 2, 3 | 982 | 3153893 |