Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081009.1 | NP_001074478.1 | 15 | 1667 | ||
GenBank mRNA | AK149889.1 | - | 14 | 1634 | ||
AK149918.1 | - | 14 | 1634 | |||
AK150208.1 | - | 14 | 1634 | |||
AK150732.1 | - | 13 | 1445 | |||
AK151510.1 | - | 13 | 1446 | |||
AK152919.1 | - | 13 | 1445 | |||
AK153169.1 | - | 13 | 1445 | |||
AK153270.1 | - | 13 | 1445 | |||
BC021315.1 | - | 5 | 801 | 5027417 | ||
BC021881.1 | - | 14 | 1637 | 3710450 | ||
EF591877.1 | - | 1402 |