Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suz12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163018.1 | NP_001156490.1 | 6 | 847 | ||
NM_199196.2 | NP_954666.1 | 7 | 916 | |||
GenBank mRNA | AK122213.1 | - | 7 | 901 | ||
BC064461.1 | - | 7 | 916 | 6821922 | ||
BC084591.1 | - | 2 | 187 | |||
W13031.1 | - | 1 | 25 |