Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166413.1 | NP_001159885.1 | 14 | 1505 | ||
NM_028027.3 | NP_082303.2 | 14 | 1567 | |||
GenBank mRNA | AF487515.1 | - | 14 | 1533 | ||
AK010452.1 | - | 14 | 1533 | |||
AK158684.1 | - | 14 | 1505 | |||
BC023367.1 | - | 5 | 544 | |||
BC041113.1 | - | 12 | 1881 | |||
BC054833.1 | - | 6 | 789 | 6439728 |