Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289774.1 | NP_001276703.1 | 11 | 1476 | ||
NM_001289775.1 | NP_001276704.1 | 11 | 1476 | |||
NM_001289776.1 | NP_001276705.1 | 12 | 1898 | |||
NM_001289777.1 | NP_001276706.1 | 13 | 1723 | |||
NM_027530.3 | NP_081806.1 | 11 | 1844 | |||
GenBank mRNA | AK013185.1 | - | 13 | 1723 | ||
AK018184.1 | - | 11 | 1822 | |||
AK083242.1 | - | 11 | 1779 | |||
AK148795.1 | - | 11 | 1140 | |||
AK154941.1 | - | 11 | 1477 | |||
AK160820.1 | - | 11 | 1478 | |||
BC017648.1 | - | 11 | 1233 | 4527858 | ||
BC049127.1 | - | 11 | 1783 | 6821567 | ||
BC058259.1 | - | 12 | 1617 | 6469137 | ||
BP765560.1 | - | 1 | 81 | |||
CT010404.1 | - | 11 | 1217 |