Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029148.1 | NP_083424.1 | 2 | 375 | ||
GenBank mRNA | AK015667.1 | - | 2 | 387 | ||
AK030696.1 | - | 2 | 389 | |||
AK033111.1 | - | 2 | 297 | |||
AK040779.1 | - | 2 | 360 | |||
AK053897.1 | - | 2 | 2506 | |||
AK129301.1 | - | 2 | 374 | |||
AK147465.1 | - | 2 | 376 | |||
BC050918.1 | - | 2 | 274 | |||
BC054558.1 | - | 2 | 375 | 6405599 | ||
BC138168.1 | - | 2 | 349 | 8861187 | ||
BC138169.1 | - | 2 | 349 | 8861188 |