Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D19Bwg1357e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050829.1 | - | 2 | 92 | ||
AK088081.1 | - | 2 | 88 | |||
AK141893.1 | - | 2 | 90 | |||
BC016186.1 | - | 2 | 105 | |||
BC041111.1 | - | 1 | 70 | |||
BC052686.1 | - | 1 | 44 | |||
BC145533.1 | - | 2 | 89 | |||
BY251590.1 | - | 2 | 179 | |||
BY289042.1 | - | 2 | 164 |