Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016771.3 | NP_058051.3 | 3, 4 | 488 | ||
GenBank mRNA | AF026073.1 | - | 3, 4 | 366 | ||
AI118913.1 | - | 500 | ||||
AK133530.1 | - | 3, 4 | 407 | |||
AK160466.1 | - | 3, 4 | 473 | |||
BC010752.1 | - | 3, 4 | 461 | |||
BC066190.1 | - | 3, 4 | 441 | 30472198 | ||
U32371.1 | - | 3, 4 | 438 |