Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016798.3 | NP_058078.1 | 4 | 512 | ||
GenBank mRNA | AF002283.1 | - | 4 | 536 | ||
AK016447.1 | - | 4 | 489 | |||
AK078421.1 | - | 4 | 512 | |||
BC070418.1 | - | 4 | 478 | 5714572 |