Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199023.1 | NP_001185952.1 | 12 | 2501 | ||
NM_001199024.1 | NP_001185953.1 | 11 | 2336 | |||
NM_001199025.1 | NP_001185954.1 | 12 | 1841 | |||
NM_016915.4 | NP_058611.1 | 11 | 1882 | |||
GenBank mRNA | AF259401.1 | - | 11 | 1781 | ||
AK144495.1 | - | 12 | 1715 | |||
AK153333.1 | - | 11 | 2142 | |||
BC003487.1 | - | 11 | 1675 | 2811357 | ||
BC049778.1 | - | 11 | 2062 | |||
BC052845.1 | - | 11 | 1707 | 30054573 | ||
BC057209.1 | - | 12 | 2094 | 5329147 | ||
U88624.2 | - | 10 | 1558 |