Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdcbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098227.1 | NP_001091697.1 | 5 | 358 | ||
NM_016807.2 | NP_058087.2 | 5 | 355 | |||
GenBank mRNA | AF003693.1 | - | 5 | 350 | ||
AF077527.1 | - | 5 | 314 | |||
AK014678.1 | - | 332 | ||||
AK028429.1 | - | 5 | 331 | |||
AK033412.1 | - | 5 | 356 | |||
AK146085.1 | - | 5 | 330 | |||
AK150278.1 | - | 5 | 323 | |||
AK150321.1 | - | 5 | 326 | |||
AK150544.1 | - | 5 | 330 | |||
AK150623.1 | - | 5 | 334 | |||
AK150661.1 | - | 5 | 323 | |||
AK150721.1 | - | 5 | 338 | |||
AK150736.1 | - | 5 | 323 | |||
AK150801.1 | - | 5 | 328 | |||
AK150996.1 | - | 5 | 330 | |||
AK151050.1 | - | 5 | 330 | |||
AK152017.1 | - | 5 | 330 | |||
AK152080.1 | - | 5 | 337 | |||
AK152109.1 | - | 5 | 330 | |||
AK152434.1 | - | 5 | 330 | |||
AK152552.1 | - | 5 | 334 | |||
AK152855.1 | - | 5 | 330 | |||
AK153112.1 | - | 5 | 334 | |||
AK159124.1 | - | 5 | 326 | |||
AK159323.1 | - | 5 | 301 | |||
AK159538.1 | - | 5 | 328 | |||
AK159677.1 | - | 5 | 337 | |||
AK165657.1 | - | 5 | 325 | |||
AK168777.1 | - | 5 | 326 | |||
AK169219.1 | - | 5 | 335 | |||
AK169687.1 | - | 5 | 330 | |||
AK171162.1 | - | 5 | 330 | |||
BC019400.1 | - | 5 | 321 | 4017442 |