Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159565.1 | NP_001153037.1 | 5, 6 | 451 | ||
NM_016890.4 | NP_058586.3 | 7 | 532 | |||
GenBank mRNA | AF098508.1 | - | 7 | 511 | ||
AK002720.1 | - | 7 | 523 | |||
AK003255.1 | - | 7 | 507 | |||
AK011840.1 | - | 7 | 529 | |||
AK159344.1 | - | 7 | 530 | |||
AK182271.1 | - | 3 | 127 | |||
AV493721.1 | - | 5, 6 | 437 | |||
BC061120.1 | - | 7 | 513 | 6774742 |