Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185153.1 | NP_001172082.1 | 6 | 798 | ||
NM_015785.2 | NP_056600.1 | 6 | 969 | |||
GenBank mRNA | AK006274.1 | - | 3 | 311 | ||
AK018889.1 | - | 3 | 313 | |||
AK018921.1 | - | 3 | 313 | |||
AK029541.1 | - | 3 | 313 | |||
BC139450.1 | - | 6 | 938 | 8862472 | ||
BC139453.1 | - | 6 | 938 | 8862475 | ||
BC145628.1 | - | 6 | 767 | 9054196 | ||
D17569.1 | - | 6 | 958 |