Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Porcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB036746.1 | - | 13 | 1409 | ||
AB036747.1 | - | 12 | 1391 | |||
AB036748.1 | - | 13 | 1406 | |||
AB036749.1 | - | 14 | 1424 | |||
AK010405.1 | - | 9 | 863 | |||
BC027292.1 | - | 12 | 2132 | |||
BC032284.1 | - | 13 | 1376 | 5362485 |