Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199044.1 | NP_001185973.1 | 16 | 3816 | ||
NM_020027.3 | NP_064411.2 | 16 | 3816 | |||
GenBank mRNA | AK156277.1 | - | 1 | 1159 | ||
AK220526.1 | - | 15 | 3596 | |||
BC052630.1 | - | 16 | 3787 | |||
BC053522.1 | - | 16 | 3792 | 6491366 |