Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276315.1 | NP_001263244.1 | 9 | 1839 | ||
NM_016912.2 | NP_058608.1 | 9 | 1573 | |||
NM_177270.4 | NP_796244.2 | 9 | 1573 | |||
GenBank mRNA | AB029065.1 | - | 9 | 1549 | ||
AB029066.1 | - | 9 | 1537 | |||
AK030598.1 | - | 9 | 1828 | |||
AK144574.1 | - | 9 | 1574 | |||
BC117894.1 | - | 9 | 1482 | 40096852 | ||
BC117895.1 | - | 9 | 1482 | 40096853 |