Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111314.1 | NP_001104784.1 | 12 | 1928 | ||
NM_019867.2 | NP_063920.2 | 10 | 1565 | |||
GenBank mRNA | AJ238898.1 | - | 10 | 1546 | ||
AK138433.1 | - | 10 | 1534 | |||
AY038025.1 | - | 10 | 1540 | |||
BC022680.1 | - | 4 | 343 | |||
BC039279.1 | - | 12 | 1928 | 4973826 |