Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020028.3 | NP_064412.2 | 2 | 273 | ||
GenBank mRNA | AK051092.1 | - | 2 | 223 | ||
AK143792.1 | - | 2 | 255 | |||
AK144861.1 | - | 2 | 274 | |||
BC060131.1 | - | 2 | 409 | 6409729 | ||
BC064676.1 | - | 2 | 409 | 6409729 |
Set Membership: |
G-protein coupled receptor |