Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020028.3 | NP_064412.2 | 3 | 1159 | ||
GenBank mRNA | AK051092.1 | - | 3 | 1109 | ||
AK144861.1 | - | 3 | 1160 | |||
BC060131.1 | - | 3 | 1295 | 6409729 | ||
BC064676.1 | - | 3 | 1295 | 6409729 |
Set Membership: |
G-protein coupled receptor |