Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172493.2 | NP_766081.1 | 21 | 2680 | ||
GenBank mRNA | AB713187.1 | - | 22 | 2475 | ||
AK083254.1 | - | 14 | 1612 | |||
AK088648.1 | - | 21 | 2614 | |||
AK153986.1 | - | 21 | 2668 | |||
AY312280.1 | - | 21 | 2617 | |||
Y15910.1 | - | 2 | 187 |