Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172493.2 | NP_766081.1 | 24 | 3139 | ||
GenBank mRNA | AB713187.1 | - | 25 | 2934 | ||
AK083254.1 | - | 17 | 2071 | |||
AK088648.1 | - | 24 | 3073 | |||
AY312280.1 | - | 24 | 3076 | |||
BB430043.2 | - | 1 | 23 | |||
Y15910.1 | - | 5 | 646 |