Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017407.2 | NP_059103.1 | 21, 22 | 3345 | ||
GenBank mRNA | AF420307.1 | - | 21, 22 | 3282 | ||
AK076619.1 | - | 21, 22 | 3343 | |||
BC052672.1 | - | 21, 22 | 3309 | 30058496 |