Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017478.3 | NP_059506.1 | 1687 | |||
GenBank mRNA | AF205065.1 | - | 1590 | |||
AJ251067.1 | - | 1659 | ||||
AK088492.1 | - | 1671 | ||||
AK147943.1 | - | 1634 | ||||
BC008136.1 | - | 212 | ||||
BC049178.1 | - | 1653 | 6309945 | |||
BC145898.1 | - | 1630 | 40131201 | |||
BC145900.1 | - | 1630 | 40131203 |