Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018740.2 | NP_061210.2 | 5 | 540 | ||
GenBank mRNA | AF004107.1 | - | 4 | 459 | ||
AK011130.1 | - | 3 | 352 | |||
AK013662.1 | - | 3 | 141 | |||
AK152582.1 | - | 5 | 544 | |||
AK199980.1 | - | 2 | 86 | |||
AK207946.1 | - | 1 | 22 | |||
BC003752.1 | - | 4 | 391 | 3489878 | ||
BY292587.1 | - | 3 | 342 |