Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019428.3 | NP_062301.3 | 4 | 262 | ||
GenBank mRNA | AK004137.1 | - | 4 | 260 | ||
AK160536.1 | - | 4 | 189 | |||
AK169253.1 | - | 4 | 189 | |||
BC062146.1 | - | 4 | 247 | 6518236 | ||
U95123.1 | - | 4 | 241 |