Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019429.2 | NP_062302.1 | 10, 11 | 1381 | ||
GenBank mRNA | AJ131775.1 | - | 10, 11 | 1381 | ||
AK040376.1 | - | 3, 4 | 764 | |||
AK088019.1 | - | 10, 11 | 1409 | |||
BC125481.1 | - | 10, 11 | 1379 | 40129996 | ||
BC131955.1 | - | 10, 11 | 1379 | 40130232 |