Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195083.1 | NP_001182012.1 | 6 | 1136 | ||
NM_001195130.1 | NP_001182059.1 | 14 | 2536 | |||
NM_018774.4 | NP_061244.1 | 14 | 2529 | |||
GenBank mRNA | AB062362.1 | - | 6 | 1122 | ||
AF060076.1 | - | 7 | 1115 | |||
AK196978.1 | - | 1 | 177 | |||
BC051657.1 | - | 6 | 1004 | |||
BC057571.1 | - | 14 | 2472 | 6410302 | ||
BC071246.1 | - | 6 | 1117 | |||
U81491.1 | - | 13 | 2426 |